siRNA Details
Gene Symbol: Gper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133573.1 | NP_598257.1 | 1 | 272 | ||
XM_006248914.2 | XP_006248976.1 | 2 | 1596 | |||
GenBank mRNA | U92802.1 | - | 1 | 272 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gper1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133573.1 | NP_598257.1 | 1 | 272 | ||
XM_006248914.2 | XP_006248976.1 | 2 | 1596 | |||
GenBank mRNA | U92802.1 | - | 1 | 272 |
Set Membership: |
![]() |