siRNA Details
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133614.2 | NP_598298.1 | 7 | 824 | ||
XM_006240114.2 | XP_006240176.1 | 7 | 2150 | |||
GenBank mRNA | AJ289714.1 | - | 7 | 668 | ||
BC089099.1 | - | 7 | 824 | 7306738 |
