siRNA Details
Gene Symbol: Slc25a21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133614.2 | NP_598298.1 | 1, 2 | 320 | ||
XM_006240114.2 | XP_006240176.1 | 1, 2 | 1646 | |||
GenBank mRNA | AJ289714.1 | - | 1, 2 | 164 | ||
BC089099.1 | - | 1, 2 | 320 | 7306738 |
