siRNA Details
Gene Symbol: Ptprg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134356.1 | NP_599183.1 | 25 | 3819 | ||
GenBank mRNA | AY177703.1 | - | 25 | 3819 | ||
AY177704.1 | - | 24 | 3732 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_008981, L09562, AK144283, BC079595.
Gene Ontology Categories:
