siRNA Details
Gene Symbol: Tmem176b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270589.1 | NP_001257518.1 | 5 | 1207 | ||
NM_001270590.1 | NP_001257519.1 | 5 | 892 | |||
NM_001270591.1 | NP_001257520.1 | 6 | 670 | |||
NM_001270592.1 | NP_001257521.1 | 6 | 667 | |||
NM_001270593.1 | NP_001257522.1 | 5 | 604 | |||
NM_001270594.1 | NP_001257523.1 | 5 | 601 | |||
NM_134390.3 | NP_599217.1 | 5 | 1210 | |||
XM_006236455.2 | XP_006236517.1 | 5 | 593 | |||
GenBank mRNA | AF370882.1 | - | 4 | 563 | ||
BC059116.1 | - | 5 | 709 | 6920949 | ||
FM070640.1 | - | 5 | 591 | |||
FQ209634.1 | - | 5 | 598 | |||
FQ212987.1 | - | 5 | 599 | |||
FQ219149.1 | - | 6 | 654 | |||
FQ219532.1 | - | 6 | 654 | |||
FQ219576.1 | - | 5 | 598 | |||
FQ220000.1 | - | 5 | 784 | |||
FQ233370.1 | - | 6 | 664 | |||
FQ235102.1 | - | 5 | 616 |
