siRNA Details
Gene Symbol: Dbp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289982.1 | NP_001276911.1 | 3 | 825 | ||
NM_001289983.1 | NP_001276912.1 | 2 | 714 | |||
NM_012543.3 | NP_036675.1 | 3 | 1125 | |||
GenBank mRNA | AY518348.1 | - | 3 | 424 | ||
AY518349.1 | - | 2 | 313 | |||
BC087668.1 | - | 3 | 1125 | 7317132 | ||
FQ213620.1 | - | 3 | 639 | |||
J03179.1 | - | 3 | 1091 |
