siRNA Details
Gene Symbol: Gspt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001003978.1 | NP_001003978.1 | 13 | 1841 | ||
GenBank mRNA | BC079092.1 | - | 13 | 1841 | 7123818 | |
FQ214757.1 | - | 4 | 356 |
Gene Symbol: LOC100911685 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003750763.3 | XP_003750811.1 | 2264 | |||
XM_006245740.2 | XP_006245802.1 | 2267 |
