siRNA Details
Gene Symbol: Ldhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012595.1 | NP_036727.1 | 2 | 117 | ||
XM_006237615.2 | XP_006237677.1 | 3 | 288 | |||
GenBank mRNA | BC059149.1 | - | 2 | 174 | 6919339 | |
FQ212899.1 | - | 2 | 180 | |||
FQ214847.1 | - | 2 | 164 | |||
FQ215687.1 | - | 2 | 180 | |||
U07181.1 | - | 2 | 117 |
Gene Symbol: Hbb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | FQ213582.1 | - | 162 |
