siRNA Details
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139194.2 | NP_631933.2 | 9 | 722 | ||
GenBank mRNA | D26113.1 | - | 9 | 747 | ||
FQ233018.1 | - | 9 | 710 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fas | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139194.2 | NP_631933.2 | 9 | 722 | ||
GenBank mRNA | D26113.1 | - | 9 | 747 | ||
FQ233018.1 | - | 9 | 710 |