siRNA Details
Gene Symbol: Arfgap1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145090.3 | NP_659558.1 | 3, 4 | 338 | ||
XM_006235731.2 | XP_006235793.1 | 3, 4 | 438 | |||
XM_006235732.1 | XP_006235794.1 | 3, 4 | 379 | |||
XM_006235733.2 | XP_006235795.1 | 2, 3 | 647 | |||
XM_006235734.2 | XP_006235796.1 | 2, 3 | 646 | |||
GenBank mRNA | BC070895.1 | - | 3, 4 | 319 | 7099840 | |
DQ167810.1 | - | 1, 2 | 55 | |||
DQ167811.1 | - | 1, 2 | 55 | |||
FQ222295.1 | - | 3, 4 | 361 | |||
FQ232032.1 | - | 3, 4 | 359 | |||
FQ233265.1 | - | 3, 4 | 359 | |||
U35776.1 | - | 3, 4 | 338 |
