siRNA Details
Gene Symbol: Tor1aip1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145092.2 | NP_659560.2 | 10 | 1455 | ||
XM_006250027.2 | XP_006250089.1 | 10 | 1000 | |||
XM_006250028.2 | XP_006250090.1 | 10 | 1586 | |||
GenBank mRNA | BC086987.1 | - | 10 | 1455 | 7129718 | |
FQ234456.1 | - | 1 | 124 | |||
U19614.1 | - | 10 | 1094 | |||
U20286.1 | - | 10 | 912 |
