siRNA Details
Gene Symbol: Ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012682.2 | NP_036814.1 | 3, 4 | 712 | ||
GenBank mRNA | BC088156.1 | - | 3, 4 | 700 | 7380759 | |
M11814.1 | - | 3, 4 | 710 | |||
X03894.1 | - | 3, 4 | 622 |
Gene Symbol: LOC100909612 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003751860.3 | XP_003751908.2 | 1, 2 | 263 |
