siRNA Details
Gene Symbol: Ucp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012682.2 | NP_036814.1 | 1, 2 | 305 | ||
GenBank mRNA | BC088156.1 | - | 1, 2 | 293 | 7380759 | |
FM087156.1 | - | 1, 2 | 304 | |||
M11814.1 | - | 1, 2 | 303 | |||
X03894.1 | - | 1, 2 | 215 |
