siRNA Details
Gene Symbol: Pdlim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017062.2 | NP_058758.1 | 5 | 638 | ||
XM_006246230.2 | XP_006246292.1 | 5 | 983 | |||
XM_006246231.2 | XP_006246293.1 | 5 | 696 | |||
GenBank mRNA | X76454.1 | - | 5 | 639 |
