siRNA Details
Gene Symbol: Pdlim4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017062.2 | NP_058758.1 | 7 | 1021 | ||
XM_006246230.2 | XP_006246292.1 | 7 | 1366 | |||
XM_006246231.2 | XP_006246293.1 | 7 | 1079 | |||
GenBank mRNA | X76454.1 | - | 7 | 1022 |
