siRNA Details
Gene Symbol: Actg2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012893.1 | NP_037025.1 | 6 | 805 | ||
XM_006236728.2 | XP_006236790.1 | 7 | 926 | |||
GenBank mRNA | BC087689.1 | - | 7 | 871 | 7310913 | |
FQ220056.1 | - | 7 | 870 | |||
FQ220823.1 | - | 7 | 871 | |||
FQ221094.1 | - | 7 | 870 | |||
FQ221892.1 | - | 7 | 870 | |||
FQ223160.1 | - | 7 | 896 | |||
FQ229904.1 | - | 7 | 870 | |||
M22323.1 | - | 6 | 805 |
