siRNA Details
Gene Symbol: Apobec1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012907.2 | NP_037039.1 | 3 | 563 | ||
XM_006237290.2 | XP_006237352.1 | 4 | 755 | |||
XM_006237291.2 | XP_006237353.1 | 4 | 811 | |||
XM_006237292.2 | XP_006237354.1 | 4 | 332 | |||
XM_006237293.2 | XP_006237355.1 | 4 | 487 | |||
XM_008763219.1 | XP_008761441.1 | 4 | 402 | |||
XM_008763220.1 | XP_008761442.1 | 4 | 271 | |||
GenBank mRNA | BC085335.1 | - | 3 | 563 | 7301703 | |
L07114.1 | - | 3 | 99 |
