siRNA Details
Gene Symbol: Faslg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_012908.1 | NP_037040.1 | 1 | 367 | ||
XM_006250148.2 | XP_006250210.1 | 4 | 1006 | |||
XM_008769607.1 | XP_008767829.1 | 5 | 1241 | |||
GenBank mRNA | U03470.1 | - | 1 | 367 |
