siRNA Details
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013087.1 | NP_037219.1 | 3, 4 | 506 | ||
GenBank mRNA | BC060583.1 | - | 3, 4 | 337 | 6918296 | |
FQ229194.1 | - | 1, 2 | 86 | |||
FQ231991.1 | - | 3, 4 | 492 | |||
FQ232886.1 | - | 3, 4 | 494 | |||
U19894.1 | - | 3, 4 | 506 |
