siRNA Details
Gene Symbol: Cd81 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013087.1 | NP_037219.1 | 7 | 854 | ||
GenBank mRNA | BC060583.1 | - | 7 | 685 | 6918296 | |
FQ229194.1 | - | 5 | 434 | |||
FQ231991.1 | - | 7 | 840 | |||
FQ232886.1 | - | 7 | 843 | |||
U19894.1 | - | 7 | 854 |
