siRNA Details
Gene Symbol: Zwint | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_147138.2 | NP_671479.1 | 3 | 297 | ||
GenBank mRNA | AF439397.1 | - | 3 | 279 | ||
AF532776.1 | - | 3 | 297 | |||
BC063144.1 | - | 3 | 289 | 6921645 | ||
EU000468.1 | - | 3 | 265 | |||
FQ228814.1 | - | 1 | 31 |
