siRNA Details
Gene Symbol: Rcan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_153724.2 | NP_714946.2 | 3, 4 | 488 | ||
XM_006248041.2 | XP_006248103.1 | 3, 4 | 688 | |||
GenBank mRNA | AB075973.1 | - | 3, 4 | 444 | ||
BC072478.1 | - | 3, 4 | 488 | 7103498 |
