siRNA Details
Gene Symbol: Nubp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009619.1 | NP_001009619.1 | 7 | 523 | ||
GenBank mRNA | BC088221.1 | - | 7 | 523 | 7321552 | |
FQ214074.1 | - | 6 | 467 | |||
FQ218444.1 | - | 7 | 536 | |||
FQ220638.1 | - | 7 | 536 | |||
FQ222868.1 | - | 7 | 540 | |||
FQ229330.1 | - | 7 | 539 |
