siRNA Details
Gene Symbol: Sar1b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001009622.1 | NP_001009622.1 | 4, 5 | 330 | ||
GenBank mRNA | BC088842.1 | - | 4, 5 | 330 | 7303357 | |
FQ215352.1 | - | 2, 3 | 165 | |||
FQ215892.1 | - | 2, 3 | 165 | |||
FQ216359.1 | - | 2, 3 | 165 | |||
FQ216811.1 | - | 2, 3 | 165 | |||
FQ217315.1 | - | 2, 3 | 165 | |||
FQ217751.1 | - | 4, 5 | 391 | |||
FQ219007.1 | - | 2, 3 | 165 | |||
FQ219108.1 | - | 2, 3 | 165 | |||
FQ219381.1 | - | 4, 5 | 392 | |||
FQ220447.1 | - | 2, 3 | 165 | |||
FQ224969.1 | - | 2, 3 | 165 | |||
FQ227071.1 | - | 2, 3 | 165 | |||
FQ233866.1 | - | 2, 3 | 165 |
