siRNA Details
Gene Symbol: Elp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001718.1 | NP_001001718.1 | 4 | 539 | ||
XM_006246639.2 | XP_006246701.1 | 5 | 733 | |||
GenBank mRNA | AY623900.1 | - | 4 | 539 | ||
BC158569.1 | - | 5 | 736 | 9101477 |
