siRNA Details
Gene Symbol: Tmem132e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134410.1 | NP_001127882.1 | 10 | 2218 | ||
XM_008767965.1 | XP_008766187.1 | 9 | 3900 | |||
GenBank mRNA | FQ211483.1 | - | 5 | 789 |
