siRNA Details
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105864.1 | NP_001099334.1 | 4 | 801 | ||
XM_006248688.2 | XP_006248750.1 | 4 | 789 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Scarf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105864.1 | NP_001099334.1 | 4 | 801 | ||
XM_006248688.2 | XP_006248750.1 | 4 | 789 |