siRNA Details
Gene Symbol: Pigz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109525.1 | NP_001102995.1 | 2, 3 | 293 | ||
GenBank mRNA | BC161994.1 | - | 2, 3 | 253 | 9034150 |
Gene Symbol: NEWGENE_1596188 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768784.1 | XP_008767006.1 | 371 |
