siRNA Details
Gene Symbol: Ficd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001010946.1 | NP_001010946.1 | 2 | 1648 | ||
XM_008769320.1 | XP_008767542.1 | 3 | 1545 | |||
XM_008769321.1 | XP_008767543.1 | 3 | 1475 | |||
GenBank mRNA | BC079197.1 | - | 2 | 1648 | 7131463 |
