siRNA Details
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008292.1 | NP_001008293.1 | 4 | 424 | ||
GenBank mRNA | BC086366.1 | - | 4 | 424 | 7305418 | |
FQ219826.1 | - | 4 | 417 |
Gene Symbol: LOC100360940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729540.3 | XP_002729586.2 | 2 | 454 | ||
XM_008764180.1 | XP_008762402.1 | 1 | 241 |
