siRNA Details
Gene Symbol: Diablo | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008292.1 | NP_001008293.1 | 1, 2 | 60 | ||
GenBank mRNA | BC086366.1 | - | 1, 2 | 60 | 7305418 | |
FQ219826.1 | - | 1, 2 | 53 |
Gene Symbol: LOC100360940 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002729540.3 | XP_002729586.2 | 1 | 90 |
