siRNA Details
Gene Symbol: Naca | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105939.2 | NP_001099409.1 | 3 | 192 | ||
NM_001198562.1 | NP_001185491.1 | 3 | 154 | |||
NM_001198580.1 | NP_001185509.1 | 3 | 5932 | |||
GenBank mRNA | BC166934.1 | - | 3 | 142 | 6922001 | |
BP494723.1 | - | 3 | 176 | |||
FQ215294.1 | - | 2 | 672 | |||
FQ221678.1 | - | 3 | 187 | |||
FQ222263.1 | - | 3 | 127 | |||
FQ225664.1 | - | 3 | 128 | |||
FQ231879.1 | - | 3 | 181 |
