siRNA Details
Gene Symbol: Il27ra | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105943.1 | NP_001099413.1 | 8 | 1000 | ||
XM_006255227.2 | XP_006255289.1 | 8 | 1024 | |||
XM_006255228.2 | XP_006255290.1 | 8 | 1026 |
