siRNA Details
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047847.1 | NP_001041312.1 | 9 | 895 | ||
GenBank mRNA | AY940050.1 | - | 9 | 895 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Wdr83 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001047847.1 | NP_001041312.1 | 9 | 895 | ||
GenBank mRNA | AY940050.1 | - | 9 | 895 |