siRNA Details
Gene Symbol: Ociad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013874.1 | NP_001013896.1 | 7, 8 | 720 | ||
XM_006250894.2 | XP_006250956.1 | 8, 9 | 766 | |||
GenBank mRNA | BC083718.1 | - | 7, 8 | 720 | 7190773 | |
FQ213030.1 | - | 7, 8 | 729 |
