siRNA Details
Gene Symbol: Upp1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001030025.1 | NP_001025196.1 | 7 | 843 | ||
XM_006251483.2 | XP_006251545.1 | 8 | 1212 | |||
XM_006251484.2 | XP_006251546.1 | 8 | 1118 | |||
XM_006251485.2 | XP_006251547.1 | 8 | 1062 | |||
XM_006251487.2 | XP_006251549.1 | 8 | 1209 | |||
XM_008770270.1 | XP_008768492.1 | 8 | 1036 | |||
GenBank mRNA | BC099752.1 | - | 7 | 843 | 5623209 |
