siRNA Details
Gene Symbol: LOC103690141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008770469.1 | XP_008768691.1 | 1, 2 | 329 |
Gene Symbol: Ccdc85a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191553.1 | NP_001178482.1 | 252 |
