siRNA Details
Gene Symbol: Chdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198731.2 | NP_942026.1 | 8 | 1740 | ||
XM_006252578.2 | XP_006252640.1 | 8 | 1805 | |||
XM_006252579.2 | XP_006252641.1 | 8 | 1759 | |||
XM_006252580.2 | XP_006252642.1 | 8 | 1565 | |||
XM_008770984.1 | XP_008769206.1 | 8 | 2108 | |||
XM_008770985.1 | XP_008769207.1 | 9 | 2197 | |||
GenBank mRNA | AY365023.1 | - | 7 | 1489 | ||
BC085787.1 | - | 8 | 1740 | 7108667 | ||
X94769.1 | - | 7 | 1015 |
