siRNA Details
Gene Symbol: Cherp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106064.1 | NP_001099534.1 | 4, 5 | 673 | ||
XM_006252796.2 | XP_006252858.1 | 4, 5 | 607 | |||
XM_006252797.2 | XP_006252859.1 | 4, 5 | 607 |
