siRNA Details
Gene Symbol: Mvb12a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199375.1 | NP_955407.1 | 3 | 295 | ||
XM_006252860.2 | XP_006252922.1 | 3 | 330 | |||
GenBank mRNA | BC061799.1 | - | 3 | 295 | 5598467 | |
FQ231372.1 | - | 3 | 332 | |||
FQ233380.1 | - | 3 | 332 | |||
FQ234388.1 | - | 3 | 332 |
