siRNA Details
Gene Symbol: Fcho1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106069.1 | NP_001099539.1 | 24 | 2214 | ||
XM_006252864.2 | XP_006252926.1 | 24 | 2581 | |||
XM_006252865.2 | XP_006252927.1 | 24 | 2585 | |||
XM_006252866.2 | XP_006252928.2 | 23 | 2159 | |||
XM_008771082.1 | XP_008769304.1 | 23 | 2159 | |||
XM_008771083.1 | XP_008769305.1 | 24 | 2235 |
