siRNA Details
Gene Symbol: Uimc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013884.1 | NP_001013906.1 | 2 | 177 | ||
XM_006253613.2 | XP_006253675.1 | 2 | 581 | |||
XM_006253616.2 | XP_006253678.1 | 2 | 580 | |||
GenBank mRNA | BC087149.1 | - | 2 | 177 | 7131093 |
