siRNA Details
Gene Symbol: Cdhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214434.8 | XP_214434.4 | 28 | 3612 | ||
GenBank mRNA | FQ211788.1 | - | 19 | 2903 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cdhr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_214434.8 | XP_214434.4 | 28 | 3612 | ||
GenBank mRNA | FQ211788.1 | - | 19 | 2903 |