siRNA Details
Gene Symbol: Ptpdc1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106104.1 | NP_001099574.1 | 7 | 1933 | ||
XM_006253700.2 | XP_006253762.1 | 6 | 2756 | |||
XM_006253701.2 | XP_006253763.1 | 7 | 2453 |
