siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145842.1 | NP_001139314.1 | 11 | 1401 | ||
NM_017113.2 | NP_058809.2 | 12 | 1635 | |||
XM_008767981.1 | XP_008766203.1 | 12 | 1716 | |||
GenBank mRNA | BC072469.1 | - | 11 | 1395 | 7098196 | |
FQ235073.1 | - | 2 | 213 | |||
M97750.1 | - | 11 | 1590 | |||
X62322.1 | - | 12 | 1609 |
