siRNA Details
Gene Symbol: Grn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017113.2 | NP_058809.2 | 11 | 1378 | ||
XM_008767981.1 | XP_008766203.1 | 11 | 1459 | |||
GenBank mRNA | M97750.1 | - | 10 | 1333 | ||
X62322.1 | - | 11 | 1352 |
