siRNA Details
Gene Symbol: LOC100909830 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772216.1 | XP_008770438.1 | 7 | 3293 |
Gene Symbol: Dcp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170469.1 | NP_001163940.1 | 9 | 1150 |
Gene Symbol: LOC100912393 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006244683.2 | XP_006244745.1 | 9 | 1163 |
Gene Symbol: LOC103690064 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008772178.1 | XP_008770400.1 | 7 | 1290 |
