siRNA Details
Gene Symbol: Sil1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_199376.1 | NP_955408.1 | 7 | 822 | ||
XM_006254542.2 | XP_006254604.1 | 7 | 856 | |||
XM_008772024.1 | XP_008770246.1 | 7 | 1144 | |||
XM_008772025.1 | XP_008770247.1 | 7 | 908 | |||
GenBank mRNA | BC062050.1 | - | 7 | 822 | 5598825 |
