siRNA Details
Gene Symbol: Vtn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019156.2 | NP_062029.2 | 6 | 919 | ||
GenBank mRNA | AY318963.1 | - | 916 | |||
BC105821.1 | - | 6 | 919 | 7371104 | ||
FQ209609.1 | - | 6 | 932 | |||
FQ209682.1 | - | 6 | 927 | |||
FQ209762.1 | - | 2 | 93 | |||
FQ210146.1 | - | 4 | 539 | |||
FQ211325.1 | - | 2 | 53 | |||
FQ212575.1 | - | 6 | 919 | |||
FQ218318.1 | - | 6 | 931 | |||
FQ218401.1 | - | 4 | 492 | |||
FQ219297.1 | - | 6 | 934 | |||
U44845.1 | - | 6 | 909 |
