siRNA Details
Gene Symbol: Irf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008722.1 | NP_001008722.1 | 5 | 571 | ||
XM_006255737.2 | XP_006255799.1 | 6 | 619 | |||
GenBank mRNA | AB120371.1 | - | 5 | 571 |
